siRNA Details
| Gene Symbol: LOC100360933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193275.1 | NP_001180204.1 | 4 | 255 | ||
| XM_008759490.1 | XP_008757712.1 | 1 | 623 | |||
| XM_008759491.1 | XP_008757713.1 | 1 | 111 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100360933| Gene Symbol: LOC100360933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193275.1 | NP_001180204.1 | 4 | 255 | ||
| XM_008759490.1 | XP_008757712.1 | 1 | 623 | |||
| XM_008759491.1 | XP_008757713.1 | 1 | 111 | |||