siRNA Details
| Gene Symbol: Btbd16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017464.2 | NP_001017464.1 | 9 | 938 | ||
| XM_006230403.2 | XP_006230465.1 | 9 | 1038 | |||
| XM_008759903.1 | XP_008758125.1 | 9 | 1038 | |||
| XM_008759904.1 | XP_008758126.1 | 7 | 704 | |||
| XM_008759905.1 | XP_008758127.1 | 9 | 1038 | |||
| XM_008759906.1 | XP_008758128.1 | 9 | 1038 | |||
| GenBank mRNA | BC079317.1 | - | 9 | 941 | 7135047 | |

Btbd16
Genomic Map