siRNA Details
| Gene Symbol: Sfxn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108527.1 | NP_001101997.1 | 3, 4 | 282 | ||
| XM_006231684.2 | XP_006231746.1 | 2, 3 | 2343 | |||
| XM_006231685.2 | XP_006231747.1 | 2, 3 | 1179 | |||
| XM_008760590.1 | XP_008758812.1 | 2, 3 | 1992 | |||
| GenBank mRNA | FQ215895.1 | - | 3, 4 | 271 | ||

Sfxn4
Genomic Map