siRNA Details
| Gene Symbol: Sfxn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108527.1 | NP_001101997.1 | 8, 9 | 500 | ||
| XM_006231684.2 | XP_006231746.1 | 7, 8 | 2561 | |||
| XM_006231685.2 | XP_006231747.1 | 7, 8 | 1397 | |||
| XM_008760590.1 | XP_008758812.1 | 6, 7 | 2295 | |||
| GenBank mRNA | FQ215895.1 | - | 8, 9 | 489 | ||

Sfxn4
Genomic Map