siRNA Details
| Gene Symbol: Znrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166300.1 | NP_001159772.1 | 4 | 411 | ||
| NM_213567.2 | NP_998732.1 | 3 | 329 | |||
| GenBank mRNA | AI232728.1 | - | 2 | 242 | ||
| FM113828.1 | - | 4 | 410 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Znrd1| Gene Symbol: Znrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166300.1 | NP_001159772.1 | 4 | 411 | ||
| NM_213567.2 | NP_998732.1 | 3 | 329 | |||
| GenBank mRNA | AI232728.1 | - | 2 | 242 | ||
| FM113828.1 | - | 4 | 410 | |||