siRNA Details
| Gene Symbol: Rab36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772925.1 | XP_008771147.1 | 6 | 1572 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab36| Gene Symbol: Rab36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772925.1 | XP_008771147.1 | 6 | 1572 | ||