siRNA Details
| Gene Symbol: Ddx21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037201.1 | NP_001032278.1 | 9 | 1478 | ||
| GenBank mRNA | BC105878.1 | - | 9 | 1478 | 7126328 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddx21| Gene Symbol: Ddx21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037201.1 | NP_001032278.1 | 9 | 1478 | ||
| GenBank mRNA | BC105878.1 | - | 9 | 1478 | 7126328 | |