siRNA Details
Gene Symbol: Pgrmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008374.1 | NP_001008375.1 | 1 | 358 | ||
GenBank mRNA | BC083571.1 | - | 1 | 358 | 7112203 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pgrmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008374.1 | NP_001008375.1 | 1 | 358 | ||
GenBank mRNA | BC083571.1 | - | 1 | 358 | 7112203 |