siRNA Details
| Gene Symbol: Pgrmc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008374.1 | NP_001008375.1 | 2 | 548 | ||
| GenBank mRNA | BC083571.1 | - | 2 | 548 | 7112203 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pgrmc2| Gene Symbol: Pgrmc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008374.1 | NP_001008375.1 | 2 | 548 | ||
| GenBank mRNA | BC083571.1 | - | 2 | 548 | 7112203 | |