siRNA Details
| Gene Symbol: Anp32e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013200.1 | NP_001013218.1 | 2 | 405 | ||
| GenBank mRNA | BC083744.1 | - | 2 | 405 | 7192147 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Anp32e| Gene Symbol: Anp32e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013200.1 | NP_001013218.1 | 2 | 405 | ||
| GenBank mRNA | BC083744.1 | - | 2 | 405 | 7192147 | |