siRNA Details
| Gene Symbol: Vangl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109584.1 | NP_001103054.1 | 4 | 781 | ||
| XM_006233054.2 | XP_006233116.1 | 4 | 822 | |||
| XM_006233055.2 | XP_006233117.1 | 4 | 821 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vangl1| Gene Symbol: Vangl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109584.1 | NP_001103054.1 | 4 | 781 | ||
| XM_006233054.2 | XP_006233116.1 | 4 | 822 | |||
| XM_006233055.2 | XP_006233117.1 | 4 | 821 | |||