siRNA Details
| Gene Symbol: LOC100911379 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233104.2 | XP_006233166.1 | 6 | 1210 | ||
| XM_008761437.1 | XP_008759659.1 | 766 | ||||
| Gene Symbol: LOC100363228 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233106.2 | XP_006233168.1 | 6 | 550 | ||

LOC100911379
Genomic Map