siRNA Details
| Gene Symbol: Gimd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113782.1 | NP_001107254.1 | 3 | 758 | ||
| XM_006233317.2 | XP_006233379.1 | 3 | 766 | |||
| XM_008761516.1 | XP_008759738.1 | 3 | 558 | |||
| GenBank mRNA | BC158622.1 | - | 3 | 758 | 7454123 | |

Gimd1
Genomic Map