siRNA Details
| Gene Symbol: Mvb12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271232.1 | NP_001258161.1 | 7 | 787 | ||
| XM_008761761.1 | XP_008759983.1 | 7 | 843 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mvb12b| Gene Symbol: Mvb12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271232.1 | NP_001258161.1 | 7 | 787 | ||
| XM_008761761.1 | XP_008759983.1 | 7 | 843 | |||