siRNA Details
| Gene Symbol: RGD1308742 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234051.2 | XP_006234113.2 | 2 | 299 | ||
| XM_008761799.1 | XP_008760021.1 | 2 | 299 | |||
| XM_008761800.1 | XP_008760022.1 | 2 | 296 | |||
| XM_342422.5 | XP_342423.5 | 2 | 296 | |||

RGD1308742
Genomic Map