siRNA Details
| Gene Symbol: Nop56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025732.1 | NP_001020903.1 | 9 | 1163 | ||
| GenBank mRNA | BC086568.1 | - | 6 | 841 | ||
| BC099149.1 | - | 9 | 1163 | 7366316 | ||
| FQ224806.1 | - | 1 | 416 | |||
| FQ232640.1 | - | 4 | 409 | |||

Nop56
Genomic Map