siRNA Details
| Gene Symbol: Nop56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025732.1 | NP_001020903.1 | 7 | 863 | ||
| GenBank mRNA | BC086568.1 | - | 4 | 541 | ||
| BC099149.1 | - | 7 | 863 | 7366316 | ||
| FQ232640.1 | - | 2 | 109 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_024193, AK154024, AK030730, AK013528, AK201684, and others.

Nop56
Genomic Map