siRNA Details
| Gene Symbol: Tbc1d20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004281.1 | NP_001004281.1 | 6 | 799 | ||
| GenBank mRNA | BC079383.1 | - | 6 | 799 | 7111239 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbc1d20| Gene Symbol: Tbc1d20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004281.1 | NP_001004281.1 | 6 | 799 | ||
| GenBank mRNA | BC079383.1 | - | 6 | 799 | 7111239 | |