siRNA Details
| Gene Symbol: Tril | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034010.1 | NP_001029182.1 | 1 | 1086 | ||
| GenBank mRNA | BC100659.1 | - | 1 | 1086 | 7935751 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tril| Gene Symbol: Tril | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034010.1 | NP_001029182.1 | 1 | 1086 | ||
| GenBank mRNA | BC100659.1 | - | 1 | 1086 | 7935751 | |