siRNA Details
| Gene Symbol: Fam13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100862.1 | NP_001094332.1 | 8 | 786 | ||
| XM_006236586.2 | XP_006236648.1 | 7 | 793 | |||
| XM_008762964.1 | XP_008761186.1 | 8 | 820 | |||
| GenBank mRNA | BC088230.1 | - | 8 | 783 | ||

Fam13a
Genomic Map