siRNA Details
| Gene Symbol: Isy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014188.1 | NP_001014210.1 | 8 | 536 | ||
| GenBank mRNA | BC079119.1 | - | 8 | 536 | 7126525 | |
| FQ218201.1 | - | 8 | 587 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Isy1| Gene Symbol: Isy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014188.1 | NP_001014210.1 | 8 | 536 | ||
| GenBank mRNA | BC079119.1 | - | 8 | 536 | 7126525 | |
| FQ218201.1 | - | 8 | 587 | |||