siRNA Details
| Gene Symbol: Rpusd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108641.1 | NP_001102111.1 | 5 | 716 | ||
| XM_006237044.2 | XP_006237106.1 | 6 | 581 | |||
| XM_006237045.2 | XP_006237107.1 | 6 | 756 | |||
| XM_006237046.2 | XP_006237108.1 | 6 | 581 | |||

Rpusd3
Genomic Map