siRNA Details
Gene Symbol: Klrb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010964.1 | NP_001010964.1 | 3 | 257 | ||
XM_006237414.2 | XP_006237476.1 | 3 | 317 | |||
GenBank mRNA | EF100677.1 | - | 3 | 241 | ||
EF100681.1 | - | 3 | 241 | |||
EF100683.1 | - | 3 | 241 | |||
FJ416340.1 | - | 3 | 262 | |||
FQ232553.1 | - | 2 | 120 | |||
M62891.1 | - | 3 | 257 |
Gene Symbol: RGD2301395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135687.1 | NP_001129159.1 | 3 | 241 | ||
GenBank mRNA | FJ158033.1 | - | 3 | 241 |
