siRNA Details
| Gene Symbol: Snapc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013212.1 | NP_001013230.1 | 6, 7 | 814 | ||
| GenBank mRNA | BC091186.1 | - | 6, 7 | 814 | 7366548 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snapc3| Gene Symbol: Snapc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013212.1 | NP_001013230.1 | 6, 7 | 814 | ||
| GenBank mRNA | BC091186.1 | - | 6, 7 | 814 | 7366548 | |