siRNA Details
| Gene Symbol: Trappc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008376.1 | NP_001008377.1 | 5 | 542 | ||
| GenBank mRNA | BC086377.1 | - | 5 | 542 | 7301917 | |
| FQ214189.1 | - | 5 | 598 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trappc3| Gene Symbol: Trappc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008376.1 | NP_001008377.1 | 5 | 542 | ||
| GenBank mRNA | BC086377.1 | - | 5 | 542 | 7301917 | |
| FQ214189.1 | - | 5 | 598 | |||