siRNA Details
| Gene Symbol: Tmem54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034151.1 | NP_001029323.1 | 4, 5 | 578 | ||
| GenBank mRNA | BC101399.1 | - | 4, 5 | 578 | 7933358 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem54| Gene Symbol: Tmem54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034151.1 | NP_001029323.1 | 4, 5 | 578 | ||
| GenBank mRNA | BC101399.1 | - | 4, 5 | 578 | 7933358 | |