siRNA Details
| Gene Symbol: Arhgef19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108692.1 | NP_001102162.1 | 4, 5 | 984 | ||
| XM_008764270.1 | XP_008762492.1 | 2, 3 | 571 | |||
| XM_008764271.1 | XP_008762493.1 | 2, 3 | 531 | |||
| XM_008764272.1 | XP_008762494.1 | 2, 3 | 1810 | |||

Arhgef19
Genomic Map