siRNA Details
| Gene Symbol: Rbks | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108703.1 | NP_001102173.1 | 8 | 873 | ||
| XM_006239805.2 | XP_006239867.1 | 7 | 754 | |||
| XM_006239806.2 | XP_006239868.1 | 7 | 730 | |||
| XM_008764520.1 | XP_008762742.1 | 8 | 1367 | |||

Rbks
Genomic Map