siRNA Details
Gene Symbol: Rrm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025740.1 | NP_001020911.1 | 2, 3 | 220 | ||
GenBank mRNA | BC099082.1 | - | 2, 3 | 220 | 7443729 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rrm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025740.1 | NP_001020911.1 | 2, 3 | 220 | ||
GenBank mRNA | BC099082.1 | - | 2, 3 | 220 | 7443729 |