siRNA Details
| Gene Symbol: Serpina11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008776.2 | NP_001008776.1 | 3 | 971 | ||
| XM_006240481.2 | XP_006240543.1 | 4 | 1076 | |||
| GenBank mRNA | AY325135.1 | - | 3 | 1011 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serpina11| Gene Symbol: Serpina11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008776.2 | NP_001008776.1 | 3 | 971 | ||
| XM_006240481.2 | XP_006240543.1 | 4 | 1076 | |||
| GenBank mRNA | AY325135.1 | - | 3 | 1011 | ||