siRNA Details
| Gene Symbol: Serpina11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008776.2 | NP_001008776.1 | 1 | 68 | ||
| NM_001166352.1 | NP_001159824.1 | 2 | 174 | |||
| XM_006240481.2 | XP_006240543.1 | 2 | 173 | |||
| GenBank mRNA | AY325135.1 | - | 1 | 108 | ||
| BC111708.1 | - | 1 | 86 | 7368690 | ||
| EX492846.1 | - | 2 | 174 | |||
| FQ211426.1 | - | 2 | 146 | |||

Serpina11
Genomic Map