siRNA Details
| Gene Symbol: Reep6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013218.1 | NP_001013236.1 | 2 | 157 | ||
| XM_006240967.2 | XP_006241029.1 | 2 | 427 | |||
| GenBank mRNA | BC083830.1 | - | 2 | 157 | 7112359 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Reep6| Gene Symbol: Reep6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013218.1 | NP_001013236.1 | 2 | 157 | ||
| XM_006240967.2 | XP_006241029.1 | 2 | 427 | |||
| GenBank mRNA | BC083830.1 | - | 2 | 157 | 7112359 | |