siRNA Details
| Gene Symbol: Ndufs7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008525.1 | NP_001008525.1 | 5 | 331 | ||
| XM_006240972.2 | XP_006241034.1 | 6 | 578 | |||
| GenBank mRNA | BC086574.1 | - | 5 | 331 | 7306868 | |
| FQ232492.1 | - | 5 | 340 | |||

Ndufs7
Genomic Map