siRNA Details
| Gene Symbol: Ckap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108740.1 | NP_001102210.1 | 2 | 633 | ||
| XM_008765245.1 | XP_008763467.1 | 2 | 1266 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ckap4| Gene Symbol: Ckap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108740.1 | NP_001102210.1 | 2 | 633 | ||
| XM_008765245.1 | XP_008763467.1 | 2 | 1266 | |||