siRNA Details
| Gene Symbol: Elk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108743.1 | NP_001102213.1 | 1 | 75 | ||
| XM_006241265.2 | XP_006241327.1 | 2 | 480 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Elk3| Gene Symbol: Elk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108743.1 | NP_001102213.1 | 1 | 75 | ||
| XM_006241265.2 | XP_006241327.1 | 2 | 480 | |||