siRNA Details
Gene Symbol: Stac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130558.1 | NP_001124030.1 | 3, 4 | 462 | ||
XM_006241486.2 | XP_006241548.1 | 3, 4 | 975 | |||
XM_006241488.2 | XP_006241550.1 | 2, 3 | 907 | |||
XM_008765415.1 | XP_008763637.1 | 3, 4 | 397 | |||
GenBank mRNA | FQ216646.1 | - | 2, 3 | 286 |
