siRNA Details
| Gene Symbol: Ptp4a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114405.2 | NP_001107877.1 | 5 | 959 | ||
| XM_006241729.2 | XP_006241791.1 | 7 | 1456 | |||
| XM_006241731.2 | XP_006241793.1 | 6 | 2566 | |||
| XM_006241732.2 | XP_006241794.1 | 6 | 1378 | |||
| XM_006241733.2 | XP_006241795.1 | 6 | 1337 | |||
| XM_008765573.1 | XP_008763795.1 | 7 | 2644 | |||
| XM_008765574.1 | XP_008763796.1 | 6 | 2023 | |||
| GenBank mRNA | BC159432.1 | - | 5 | 959 | 9092581 | |

Ptp4a3
Genomic Map