siRNA Details
Gene Symbol: Ptp4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114405.2 | NP_001107877.1 | 5 | 998 | ||
XM_006241729.2 | XP_006241791.1 | 7 | 1495 | |||
XM_006241731.2 | XP_006241793.1 | 6 | 2605 | |||
XM_006241732.2 | XP_006241794.1 | 6 | 1417 | |||
XM_006241733.2 | XP_006241795.1 | 6 | 1376 | |||
XM_008765573.1 | XP_008763795.1 | 7 | 2683 | |||
XM_008765574.1 | XP_008763796.1 | 6 | 2062 | |||
GenBank mRNA | BC159432.1 | - | 5 | 998 | 9092581 |
