siRNA Details
Gene Symbol: Ptp4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114405.2 | NP_001107877.1 | 2, 3 | 687 | ||
XM_006241729.2 | XP_006241791.1 | 4, 5 | 1184 | |||
XM_006241731.2 | XP_006241793.1 | 3, 4 | 2294 | |||
XM_006241732.2 | XP_006241794.1 | 3, 4 | 1106 | |||
XM_006241733.2 | XP_006241795.1 | 3, 4 | 1065 | |||
XM_008765573.1 | XP_008763795.1 | 4, 5 | 2372 | |||
XM_008765574.1 | XP_008763796.1 | 3, 4 | 1751 | |||
GenBank mRNA | BC159432.1 | - | 2, 3 | 687 | 9092581 |
