siRNA Details
| Gene Symbol: Lrrc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135896.1 | NP_001129368.1 | 6 | 1608 | ||
| XM_008765581.1 | XP_008763803.1 | 4 | 1426 | |||
| XM_008765582.1 | XP_008763804.1 | 4 | 1445 | |||
| XM_008765583.1 | XP_008763805.1 | 5 | 2149 | |||
| GenBank mRNA | BC127521.1 | - | 6 | 1608 | 7110443 | |

Lrrc24
Genomic Map