siRNA Details
| Gene Symbol: Chadl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765807.1 | XP_008764029.1 | 2 | 694 | ||
| XM_008765808.1 | XP_008764030.1 | 2 | 694 | |||
| XM_008765809.1 | XP_008764031.1 | 3 | 2034 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chadl| Gene Symbol: Chadl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765807.1 | XP_008764029.1 | 2 | 694 | ||
| XM_008765808.1 | XP_008764030.1 | 2 | 694 | |||
| XM_008765809.1 | XP_008764031.1 | 3 | 2034 | |||