siRNA Details
| Gene Symbol: Rnf214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750501.3 | XP_003750549.1 | 13 | 1946 | ||
| XM_008766183.1 | XP_008764405.1 | 14 | 2051 | |||
| XM_008766184.1 | XP_008764406.1 | 14 | 2089 | |||
| XM_008766185.1 | XP_008764407.1 | 14 | 1995 | |||
| XM_008766186.1 | XP_008764408.1 | 14 | 1994 | |||
| XM_008766187.1 | XP_008764409.1 | 14 | 1586 | |||

Rnf214
Genomic Map