siRNA Details
Gene Symbol: Rnf214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750501.3 | XP_003750549.1 | 3 | 244 | ||
XM_008766183.1 | XP_008764405.1 | 3 | 244 | |||
XM_008766184.1 | XP_008764406.1 | 3 | 282 | |||
XM_008766185.1 | XP_008764407.1 | 3 | 188 | |||
XM_008766186.1 | XP_008764408.1 | 3 | 187 |
