siRNA Details
| Gene Symbol: RGD1305645 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271051.1 | NP_001257980.1 | 4 | 629 | ||
| GenBank mRNA | BG662789.1 | - | 4 | 378 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1305645| Gene Symbol: RGD1305645 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271051.1 | NP_001257980.1 | 4 | 629 | ||
| GenBank mRNA | BG662789.1 | - | 4 | 378 | ||