siRNA Details
| Gene Symbol: Atg16l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108809.3 | NP_001102279.2 | 15 | 1476 | ||
| XM_006245419.2 | XP_006245481.1 | 16 | 1702 | |||
| XM_006245420.2 | XP_006245482.1 | 14 | 1596 | |||
| GenBank mRNA | CK481169.1 | - | 3 | 195 | ||
| EV774911.1 | - | 9 | 770 | |||

Atg16l1
Genomic Map