siRNA Details
| Gene Symbol: Traf3ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012204.1 | NP_001012204.1 | 3 | 328 | ||
| XM_006245435.2 | XP_006245497.2 | 3 | 335 | |||
| XM_006245436.2 | XP_006245498.2 | 3 | 334 | |||
| XM_006245437.2 | XP_006245499.2 | 3 | 337 | |||
| GenBank mRNA | BC083645.1 | - | 3 | 328 | 7135023 | |

Traf3ip1
Genomic Map