siRNA Details
| Gene Symbol: Cav2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_131914.2 | NP_571989.2 | 1, 2 | 279 | ||
| GenBank mRNA | AF439780.1 | - | 1, 2 | 156 | ||
| BC062059.1 | - | 1, 2 | 279 | 5598269 | ||
| FQ209775.1 | - | 1, 2 | 333 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cav2| Gene Symbol: Cav2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_131914.2 | NP_571989.2 | 1, 2 | 279 | ||
| GenBank mRNA | AF439780.1 | - | 1, 2 | 156 | ||
| BC062059.1 | - | 1, 2 | 279 | 5598269 | ||
| FQ209775.1 | - | 1, 2 | 333 | |||