siRNA Details
| Gene Symbol: Prrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191888.1 | NP_001178817.1 | 5 | 403 | ||
| XM_006256963.2 | XP_006257025.1 | 6 | 567 | |||
| XM_006256965.2 | XP_006257027.1 | 4 | 397 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prrg1| Gene Symbol: Prrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191888.1 | NP_001178817.1 | 5 | 403 | ||
| XM_006256963.2 | XP_006257025.1 | 6 | 567 | |||
| XM_006256965.2 | XP_006257027.1 | 4 | 397 | |||