siRNA Details
| Gene Symbol: Snx12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108817.2 | NP_001102287.2 | 3 | 392 | ||
| XM_006257078.1 | XP_006257140.1 | 3 | 461 | |||
| XM_006257079.1 | XP_006257141.1 | 3 | 461 | |||
| GenBank mRNA | BP500254.1 | - | 3 | 398 | ||

Snx12
Genomic Map