siRNA Details
Gene Symbol: Snx12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108817.2 | NP_001102287.2 | 1, 2 | 222 | ||
XM_006257078.1 | XP_006257140.1 | 1, 2 | 291 | |||
XM_006257079.1 | XP_006257141.1 | 1, 2 | 291 | |||
GenBank mRNA | BP500254.1 | - | 1, 2 | 228 |
